missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ PV350
Click to view available options
Size: Large, 10
Description
- Rugged PVC coating for good abrasion resistance over a cotton/jersey liner for enhanced comfort and flexibility
- Rough grip, combined with a 14 gauntlet
- Provides excellentwet grip for more secure handling
- CE CAT III - EN ISO 374-1:2016/TYPE B, AKLMP
- EN388: 2016 3111X - EN ISO 374-5:2016
- Food contact compatible
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Certifications/Compliance (Europe) | CE Cat III |
Color | Black |
Exterior Finish | Rough |
Latex Free | No |
Material | PVC |
Size | Large, 10 |
Style | Gauntlet / Anatomical shape |
Thickness (English) | 55 mil |
Thickness (Metric) | 1.40 mm |
EN388 Standard | EN388: 2016 3111X |
Show More |
- Store away from light and humidity
- Rinse gloves in running water before removing, using a neutral detergent if necessary
- Gloves should not be used around heat, open flame, sparks or in potentially flammable environments
- For Professional Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'