Learn More
Masterflex™ Tygon™ E-3603 Tubing
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ E-3603 Tubing handles virtually all inorganic chemicals found in the lab.
£57.55 - £473.00
Specifications
Certifications/Compliance | FDA, NSF, 3-A, USP Class VI, Japan Food Sanitation Law #370/1959, 1935/2004/EC, 10/2011/EU |
---|---|
Autoclavable | Autoclavable |
For Use With (Application) | For laboratory, food and, beverage and vacuum applications |
Packaging Quantity | 1 Ea. |
Color | Clear |
Product Code | Brand | Diameter (Metric) Inner | Diameter (Metric) Outer | Thickness (Metric) | Overall Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Diameter (Metric) Outer | Thickness (Metric) | Overall Length (Metric) | Price | Quantity & Availability | |||||
15197064
|
Masterflex™
ACF1S1507-C |
10 mm | 14 mm | 2.4 mm | 15 m |
£240.00
15m |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
15147064
|
Masterflex™
ACF1S1518 |
4 mm | 6 mm | 1 mm | 15 m |
£82.00
15m |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||||
10323642
|
Masterflex™
ACF1S1518-C |
4 mm | 6 mm | 1 mm | 15.2 m |
£67.50
15m |
In Stock
Log in to see stock. ![]() |
|||||
15127064
|
Masterflex™
ACF1S1502 |
2 mm | 4 mm | 1 mm | 15 m |
£57.55
15m |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||||
10170161
|
Masterflex™
ACF00010-C |
4 mm | 7.1 mm | 1.6 mm | 15 m |
£123.60
15m |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
10256294
|
Masterflex™
ACF1S1503-C |
5 mm | 8 mm | 1.5 mm | 15.2 m |
£191.46
15m |
Estimated Shipment: 24-04-2025
Log in to see stock. |
|||||
12365889
|
Masterflex™
ACF00019 |
6.4 mm | 12.7 mm | 3.2 mm | 15 m |
£473.00
15m |
Estimated Shipment: 02-06-2025
Log in to see stock. |
|||||
15157064
|
Masterflex™
ACF1S1503 |
5 mm | 8 mm | 1.5 mm | 15 m |
£94.78
15m |
Estimated Shipment: 31-03-2025
Log in to see stock. |
|||||
15137064
|
Masterflex™
ACF1S1517 |
3 mm | 5 mm | 1 mm | 15 m |
£76.70
15m |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
15187064
|
Masterflex™
ACF1S1506 |
8 mm | 12 mm | 2 mm | 15 m |
£309.00
15m |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
10240053
|
Masterflex™
ACF1S1504-C |
6 mm | 9 mm | 1.5 mm | 15 m | N/A | ||||||
15167064
|
Masterflex™
ACF1S1504 |
6 mm | 9 mm | 2.4 mm | 15 m |
£144.20
15m |
Estimated Shipment: 27-06-2025
Log in to see stock. |
|||||
Description
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Saint Gobain Performance Plastics™ Tygon E-3603 Tubing handles virtually all inorganic chemicals found in the lab.
Tygon™ E-3603 tubing from Saint-Gobain is crystal clear and flexible, handling virtually all organic chemicals found in the modern laboratory. It is non-oxidizing and non-contaminating, making it an ideal choice for a variety of applications. Tygon™ E-3603 has high tolerances and lot-to-lot consistency, which will yield reproducible results in your laboratory. It is non-toxic and non-contaminating, and less permeable than rubber tubing. The glassy-smooth inner bore helps prevent buildup to facilitate cleaning.
Tygon™ E-3603 tubing is specially formulated for resistance to flex-fatigue and abrasion. As a tubing for instrumentation connection, vent, drain and other general laboratory applications, Tygon™ E-3603 tubing offers superior life, which minimizes the labor and the expenses associated with replacement.
Specifications
FDA, NSF, 3-A, USP Class VI, Japan Food Sanitation Law #370/1959, 1935/2004/EC, 10/2011/EU | |
For laboratory, food and, beverage and vacuum applications | |
Clear |
Autoclavable | |
1 Ea. | |
Flexible |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.