missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences™ Tygon™ F-4040-A Fuel and Lubricant Tubing

Lubricant and Fuel F-4040-A Tubing resists the swelling and hardening caused by hydrocarbon-based fluids, significantly reducing the risk of failure due to cracks and leaks.

Brand:  Saint Gobain Life Sciences™ AAG00017

 View more versions of this product

Product Code. 17923451

  • £111.00 / 15m

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Minimum extractability safeguards the liquid or vapor being transferred against adulteration
  • Tubing is routinely used to handle gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants
  • Very flexible and ideal for small engines due to its ability to maneuver tight places and simplify installation
  • Translucent yellow color stands out for identification purposes, yet allows you to easily view and monitor the fuels and lubricants flowing through it
  • Compatible with most hydrocarbons
  • Specifically developed for fuels and lubricants
  • Ozone- and UV- resistant
  • Applications include: small engine fuel lines, general automotive, recreational vehicles, lawn and garden equipment, coolant transfer, heating fuels, cutting compounds, polishing equipment, lubrication lines
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tubing
0.252 in.
Thermoplastic
9.6 mm
15 m
1.59 mm
167 lbf/in.
50 psi
Not Autoclavable
6.4 mm
0.378 in.
49.2 in.
0.063 in.
Fuel and Lubricant Lines, Small Engines
Yellow
29.9 in. Hg (at 73°F)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.