missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Tygon™ S3™ E-LFL 2-Stop Pump Tubing

Fits tightly to glass and metal, bends to small radius curves, is nontoxic, and transparent. Saint Gobain Life Sciences Tygon™ S3™ E-LFL 2-Stop Pump Tubing has the longest flexible life of all Tygon™ clear tubing.

Brand:  Saint Gobain Life Sciences 070602-18-ND

Code : 02

Additional Details : Weight : 0.80000kg

Product Code. 15121733

  • £109.00 / Pack of 12
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

  • Longest flex life of any clear Tygon™ tubing, reduces downtime due to pump failure
  • Broad chemical resistance
  • Excellent for laboratory applicationsCan be autoclave
  • Suitable for high purity applications
  • Extremely low particle spallation
  • Reliable for sensitive-fluid applications
  • Meets USP Class VI, ISO 10993 and FDA criteria
  • Contains no BPA or phthalate
Specifications

Specifications

USP Class VI, ISO 10993, NSF-51, FDA criteria
2.79 mm
4.48 mm
12 Pack
56 (Shore A)
Violet/White
Tygon LFL
0.85 mm
Clear
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers