Learn More
Heidolph instruments Tygon™ (Standard) Tubing
For standard applications. For Single- and Multi-Channel use (not suitable with cassette small).
Brand:  Heidolph instruments 525-69000-00
Additional Details : Weight : 0.08000kg
 View more versions of this product
Product Code. 10585492
  • £54.00 / Each
Estimated Shipment: 24-09-2024
to see stock.
Add to basket
Description
Description
  • Non-toxic, non-oxidizing.
  • Good resistance to acids, bases and inorganic media
  • Very low gas permeability, good performance life
Specifications
Specifications
Autoclavable
Borosilicate Glass
2.5 mm
Continuous/short time 0.5/1.5 bar
6.3 mm
11.3 mm
Non-sterile