missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Type 145 Cellulose Triacetate (CTA) Ultrafiltration Membranes

Type 145 Cellulose Triacetate (CTA) Ultrafiltration Membranes are highly hydrophilic with very low non-specific adsorption. Cast without any membrane support they are ideal for sample cleaning and protein removal, as well as for applications where high recovery from the ultrafiltrate is required.

Brand:  Sartorius™ 14549--47------N

1262.38 GBP valid until 2024-12-31
Use promo code "23270" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10140092

  • £1465.00 / Pack of 100
Estimated Shipment: 17-09-2024
to see stock.
Add to basket

Description

Description

  • Type: Ultrafiltration Membrane
  • Material: Cellulose Triacetate (CTA)
  • Grade: Type 145
  • Shape: Disc
  • Flow Rate: 0.11 mL/min./cm2
  • Thickness: 120 μm
  • For Use With (Application): Concentration, Protein Removal, Desalting, Buffer Exchange, and Electro-Ultrafiltration (in BioPharma, Research, and Environmental Fields)
Specifications

Specifications

Membrane Filter
Cellulose Triacetate
Non-sterile
Concentration, Protein Removal, Desalting, Buffer Exchange, Electro-Ultrafiltration
Ultra-filtration Membrane
100 Pk.
120 μm
47 mm
Ultrafiltration Disc
20 kDa
0.11 mL/min./cm2
Type 145
100 Pk.
Disc
Ultrafiltration Membrane
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers