Learn More
Sartorius™ Type 145 Cellulose Triacetate (CTA) Ultrafiltration Membranes
Type 145 Cellulose Triacetate (CTA) Ultrafiltration Membranes are highly hydrophilic with very low non-specific adsorption. Cast without any membrane support they are ideal for sample cleaning and protein removal, as well as for applications where high recovery from the ultrafiltrate is required.
Brand: Sartorius™ 14549--25------D
96.66 GBP valid until 2024-12-31
Use promo code "23270" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- Type: Ultrafiltration Membrane
- Material: Cellulose Triacetate (CTA)
- Grade: Type 145
- Shape: Disc
- Flow Rate: 0.11 mL/min./cm2
- Thickness: 120 μm
- For Use With (Application): Concentration, Protein Removal, Desalting, Buffer Exchange, and Electro-Ultrafiltration (in BioPharma, Research, and Environmental Fields)
Specifications
Membrane Filter | |
Cellulose Triacetate | |
Non-sterile | |
Concentration, Protein Removal, Desalting, Buffer Exchange, Electro-Ultrafiltration | |
Ultra-filtration Membrane | |
10 Pk. | |
120 μm | |
25 mm |
Ultrafiltration Disc | |
20 kDa | |
0.11 mL/min./cm2 | |
Type 145 | |
10 Pk. | |
Disc | |
Ultrafiltration Membrane |