missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 30 Combi Safety Storage Cabinets K-PHOENIX-30
Safe and approved storage of aggressive and flammable hazardous substances in working areas
£3218.69 - £3402.07
Specifications
Certifications/Compliance | Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 598 mm |
Depth (Metric) Interior | 519 mm |
Color | Light Gray |
Dimensions (D x W x H) | 598 x 1202 x 1968 mm |
Product Code | Brand | No. of Shelves | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Shelves | Price | Quantity & Availability | |||||
15924326
|
asecos™
3034200130345 |
9 |
£3402.07
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13264869
|
asecos™
3034200130344 |
7 |
£3218.69
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Two in one: 2 compartments (vertical seperated) allow the storage of flammable liquids and aggressive chemicals in one cabinet
- User-friendly and safe (folding door compartment): door open arrest system (standard), convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible) / cylinder lock
Specifications
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
519 mm | |
598 x 1202 x 1968 mm | |
Combi safety storage cabinets | |
Safe and approved storage of aggressive and flammable hazardous substances in working areas | |
1691 mm | |
K30.197.120.MV.FWAS | |
Powder-coated plain | |
532 mm |
598 mm | |
Light Gray | |
Steel | |
Wing, Folding | |
1968 mm | |
Door open arrest system | |
2 | |
1202 mm |