missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 30 Safety Storage Cabinet Q-CLASSIC-30 with 2 Doors

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3100306130003

Code : NEW

Additional Details : Weight : 222.00000kg

 View more versions of this product

Product Code. 15430204

  • £2750.70 / Each
Estimated Shipment: 26-11-2024
to see stock.

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Easy handling with comfort: doors stop in any position, self-closing in the event of fire
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing  locking system possible)
  • Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications

Specifications

1626 mm
Safety storage cabinet
1050 mm
Powder-coated textured
1164 mm
271.50 kg
Gray
620 mm
Wing
3
2
3 x shelf, 1 x perforated insert (375 mm), 1 x bottom collecting sump (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
519 mm
Steel
620 x 1164 x 1947 mm
519 x 1050 x 1626 mm
Standard
1947 mm
Q30.195.116
90 mm
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers