missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 30 Safety Storage Cabinet Q-CLASSIC-30 Self Closing
Safe and approved storage of flammable hazardous substances in working areas
£1667.60 - £2249.44
Specifications
Certifications/Compliance | Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant, FM 6050 approved, UL/ULC 1275 listed |
---|---|
Configuration | 3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) |
Depth (Metric) Exterior | 620 mm |
Depth (Metric) Interior | 519 mm |
Height (Metric) Opening | 90 mm |
Alert:
For indoor use only
Product Code | Brand | Width (Metric) Exterior | Door Style | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) Exterior | Door Style | Color | Price | Quantity & Availability | |||
15917706
|
asecos™
34645-061-34617 |
564 mm | Wing, Right-hinged, Self-closing | Anthracite Gray, Light Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15917716
|
asecos™
34653-060-34628 |
864 mm | Wing, Right-hinged, Self-closing | Anthracite Gray, Warning Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15937696
|
asecos™
34641-061-34617 |
564 mm | Wing, Left-hinged, Self-closing | Anthracite Gray, Light Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15937706
|
asecos™
34649-060-34628 |
864 mm | Wing, Left-hinged, Self-closing | Anthracite Gray, Warning Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15937726
|
asecos™
34659-061-34637 |
1164 mm | Wing, Self-closing | Anthracite Gray, Light Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15957716
|
asecos™
34653-061-34628 |
864 mm | Wing, Right-hinged, Self-closing | Anthracite Gray, Light Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15977696
|
asecos™
34645-060-34617 |
564 mm | Wing, Right-hinged, Self-closing | Anthracite Gray, Warning Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15977706
|
asecos™
34649-061-34628 |
864 mm | Wing, Left-hinged, Self-closing | Anthracite Gray, Light Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15997686
|
asecos™
34641-060-34617 |
564 mm | Wing, Left-hinged, Self-closing | Gray, Anthracite Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15997716
|
asecos™
34659-060-34637 |
1164 mm | Wing, Self-closing | Anthracite Gray, Warning Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Easy handling with comfort: door stops in any position, self-closing in the event of fire
- Easy handling with comfort: smooth doors with permanent self-closing function and oil-dampened door closer; open doors with a minimum amout of force
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Triple certified: cabinet tested in accordance with EN 14470-1, FM Standard 6050 and UL/ULC Standard 1275
Specifications
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant, FM 6050 approved, UL/ULC 1275 listed | |
620 mm | |
90 mm | |
Safety storage cabinet | |
1947 mm | |
3 | |
Powder-coated textured |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | |
519 mm | |
Steel | |
Safe and approved storage of flammable hazardous substances in working areas | |
1626 mm | |
Standard |