missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Type 5/6 Classic Chemical Protection Suit
247.08 GBP valid until 2024-12-31
Use promo code "23255" to get your promotional price.
Use promo code "23255" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Lightweight and breathable material guarantees an excellent combination of comfort and safety
- Particle-tight and spray-tight to a limited extent
- Free of substances that interfere with wetting agents
- Material: Polypropylene non woven laminated with Polyethylene film
- Self-adhesive zipper flap for optimal protection
- Anti-static properties
- Protection against infectious diseases
- Skin-friendly non-woven material inside
- Elasticated waistband for a perfect fit
- Secure and convenient closures thanks to elasticated bands on hood, arms and legs
- Two-way zipper
Specifications
Product Type | Cleanroom Coveralls |
Size | Small |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
Certifications/Compliance | EN 14126, ISO 13982-1 - type 5, EN 13034 - type 6, EN 1149-5, EN 1073-2 |
For Use With (Application) | Controlled Environments |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'