missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Type 5/6 Classic Chemical Protection Suit with Feet
268.76 GBP valid until 2024-12-31
Use promo code "23255" to get your promotional price.
Use promo code "23255" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- PP spunbond non-woven fabric with PE film laminated
- Extremely lightweight and breathable material guarantees an outstanding combination of comfort and safety
- Soft non-woven material lining
- Inner overlock seam for outstanding protection against particles and liquid splashes
- Middle finger loops prevent the sleeve from riding up
- Optimum fit for a high level of wearer comfort
- Safe and comfortable hems thanks to elasticated bands in the hood and sleeves
- Integrated boot sock
- Particle-tight and limited spray-tightness
- Anti-static properties
- Protection from infectious agents
- Free from paint-wetting impairement substances
Specifications
Product Type | Cleanroom Coveralls |
Size | Small |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
Certifications/Compliance | Chemical protection/PPE category III, EN ISO 13982-1:2004+A1:2010 (type 5), EN 13034:2005+A1:2010 (type 6), EN 1149-5:2008, EN 14126:2003+AC:2004, EN 1073-2:2002 |
For Use With (Application) | Controlled Environments |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'