missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Recirculating Air Filter Storage Cabinet
Recirculating air filter storage cabinet
Brand: asecos™ 32933-050-30805
Additional Details : Weight : 180.00000kg
Alert:
For indoor use only
Description
- Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface
- All in one: different compartments (multiple seperation) allow the storage of different hazardous materials in one cabinet
- No unauthorised use: doors lockable with cylinder locking (integrated in the turning handle), each compartment can be opened and locked separately
- Easy alignment: adjusting aids to compensate for uneven floor
- Ideal for the direct installation in working areas – very low noise, only approx. 39 dB
- Monitoring electronics for extraction incl. cool-device cable and power plug
- Optical and acoustical alarm including potential-free alarm contact
- Display with innovative touch panel
- Manual check of the filter saturation
- DIN EN 16121/16122 conformity
- CE compliant
Specifications
Recirculating air filter storage cabinet CX-CLASSIC-MultiRisk model CX.229.105.MM.4WD body colour anthracite gray (RAL 7016), with wing doors in Pure white (RAL 9010), interior equipment with 4 x shelf, 2 x bottom collecting sump, 4 x perforated insert, 2 x bottom collecting sump (sheet steel galvanised) | |
Safe, approved and separated storage of water pollutants, non-flammable liquids and toxics in working areas | |
41.54 in. | |
Metal | |
Cabinet and interior equipment | |
20.47 in. | |
White | |
520 mm | |
50 kg | |
CX.229.105.MM.4WD | |
4 | |
4 |
Recirculating air filter storage cabinet | |
90.16 in. | |
Textured | |
1055 mm | |
520 x 1055 x 2290 mm | |
493 x 996 x 1835 mm | |
Shelf | |
2290 mm | |
Wing | |
1055 mm | |
1835 mm | |
DIN EN 16121/16122 conformity, CE compliant |