missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Recirculating Air Filter Storage Cabinet
Recirculating air filter storage cabinet
£5506.70 - £7455.00
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant |
---|---|
Depth (Metric) | 615 mm |
Depth (Metric) Exterior | 615 mm |
Color | Warning Yellow |
Depth (English) Exterior | 24.21 in. |
Alert:
For indoor use only
Product Code | Brand | Model | Dimensions (D x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Dimensions (D x W x H) | Price | Quantity & Availability | ||||
17719989
|
asecos™
32910-040-33622 |
FX90.229.090.WDAC | 615 x 893 x 2294 mm |
£5506.70
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17774079
|
asecos™
32915-040-33600 |
FX90.229.060.WDAC | 615 x 599 x 2294 mm |
£7455.00
Each |
Estimated Shipment: 17-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Effortless use: one-hand operation for double-door cabinets (AGT)
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed: serial automatic door closing (TSA) releases arrested doors after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorized use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ideal for the direct installation in working areas – very low noise, only approx. 39 dB (A)
- With multi-stage wideband filter
- Monitoring electronics for extraction and filtration incl. cool-device cable and power plug
- Optical and acoustical alarm including potential-free alarm contact
- Display with innovative touch panel
- Fire resistance 90 minutes
- Type tested in accordance with EN 14470-1
- DIN EN 16121/16122 conformity
- CE compliant
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant | |
615 mm | |
24.21 in. | |
1647 mm | |
Recirculating air filter storage cabinet | |
Safe and approved storage of flammable liquids and further hazardous substances in working areas | |
Cabinet and interior equipment | |
3 | |
Textured |
615 mm | |
Warning Yellow | |
90.31 in. | |
Gypsum | |
Wing, Auto-close | |
2294 mm | |
75 kg | |
Shelf |