missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos Type 90 Safety Storage Cabinet Q-CLASSIC-90, Self-closing Doors
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos 38074-049-38102
2950.51 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 482.50000kg
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Easy handling with comfort: smooth doors with permanent self-closing function and oil-dampened door closer; open doors with a minimum amount of force
- No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy transport: integrated transport base with removable, optional base cover
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
Certificates
- Fire resistance: 90 minutes
- Type tested in accordance with EN 14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
1050 mm | |
76.89 in. | |
Gypsum | |
615 x 1193 x 1953 mm | |
24.21 in. | |
1050 x 522 x 1647 mm | |
1035 lb. | |
4 | |
64.84 in. | |
Safety Storage Cabinet | |
46.97 in. | |
Silver | |
1953 mm | |
Q90.195.120.ME.WDC | |
2 |
522 mm | |
Textured | |
(4) grid (steel galvanized), (1) bottom collecting sump (sheet steel powder-coated) | |
470 kg | |
20.55 in. | |
615 mm | |
120 kg | |
41.34 in. | |
1647 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
1193 mm | |
Grid | |
Self-close | |
360/360/360/370 mm | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant |