missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet Q-CLASSIC-90 Single Door
Safe and approved storage of flammable hazardous substances in working areas
£2556.72 - £3187.42
Specifications
Depth (Metric) Exterior | 615 mm |
---|---|
Depth (Metric) Interior | 522 mm |
Dimensions (D x W x H) | 615 x 599 x 1953 mm |
Dimensions (D x W x H) Interior | 522 x 450 x 1647 mm |
Height (Metric) Opening | 90 mm |
Product Code | Brand | Configuration | Door Style | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Door Style | Description | Price | Quantity & Availability | |||
15941596
|
asecos™
3007304034371 |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Swing, Right-hinged | Warning Yellow doors |
£2556.72
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15931396
|
asecos™
3007204133600 |
3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Swing, Left-hinged | Light gray doors |
£2556.72
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15961636
|
asecos™
3007304134371 |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Swing, Right-hinged | Light gray doors |
£2556.72
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15981596
|
asecos™
3007304034375 |
3 x tray shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Swing, Right-hinged | Warning Yellow doors |
£2758.12
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15971366
|
asecos™
3007204033606 |
3 x tray shelf, 1 x perforated insert (180 mm high), 1 x bottom collecting sump (sheet steel powder-coated) | Swing, Left-hinged | Warning Yellow doors |
£2758.12
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15991596
|
asecos™
3007304034376 |
4 x drawer (sheet steel powder-coated) | Swing, Right-hinged | Warning Yellow doors |
£2907.58
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15981396
|
asecos™
3007204133608 |
4 x drawer (sheet steel powder-coated) | Swing, Left-hinged | Light gray doors |
£2907.58
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15911646
|
asecos™
3007304134376 |
4 x drawer (sheet steel powder-coated) | Swing, Right-hinged | Light gray doors |
£2907.58
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15997822
|
asecos™
3007204033610 |
6 x drawer (sheet steel powder-coated) | Swing, Left-hinged | Warning yellow doors |
£3187.42
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15911606
|
asecos™
3007304034378 |
6 x drawer (sheet steel powder-coated) | Swing, Right-hinged | Warning Yellow doors |
£3187.42
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15901406
|
asecos™
3007204133610 |
6 x drawer (sheet steel powder-coated) | Swing, Left-hinged | Light gray doors |
£3187.42
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15931646
|
asecos™
3007304134378 |
6 x drawer (sheet steel powder-coated) | Swing, Right-hinged | Light gray doors |
£3187.42
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Safe and approved storage of flammable hazardous substances in working areas
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Easy handling with comfort: doors stop in any position, self-closing in the event of fire
- No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy transport: integrated transport base with removable, optional base cover
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
- Fire resistance 90 minutes
- Type tested in accordance with EN 14470-1
- EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
615 mm | |
615 x 599 x 1953 mm | |
90 mm | |
Safety storage cabinet | |
1953 mm | |
1 | |
599 mm |
522 mm | |
522 x 450 x 1647 mm | |
Steel | |
Safe and approved storage of flammable hazardous substances in working areas | |
1647 mm | |
Powder-coated textured | |
450 mm |