missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet Q-CLASSIC-90 Winged Doors
Safe and approved storage of flammable hazardous substances in working areas
£4129.23 - £4759.93
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 615 mm |
Depth (Metric) Interior | 522 mm |
Dimensions (D x W x H) | 615 x 1193 x 1953 mm |
Dimensions (D x W x H) Interior | Left: 522 x 450 x 1647 mm, Right: 522 x 450 x 1647 mm |
Alert:
For indoor use only
Product Code | Brand | Description | No. of Doors | No. of Shelves | Weight (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | No. of Doors | No. of Shelves | Weight (Metric) | Price | Quantity & Availability | ||
15967616
|
asecos™
3421404133657 |
Light gray doors | 2 | 3 | 528.50 kg |
£4759.93
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15987606
|
asecos™
3421404033656 |
Warning Yellow doors | 2 | 6 | 504.50 kg |
£4129.23
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15997606
|
asecos™
3421404033657 |
Warning Yellow doors | 2 | 3 | 528.50 kg |
£4759.93
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- Safe and approved storage of flammable hazardous substances in working areas
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Easy handling with comfort: doors stop in any position, self-closing in the event of fire
- No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy transport: integrated transport base with removable, optional base cover
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
- Fire resistance 90 minutes
- Type tested in accordance with EN 14470-1
- EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
522 mm | |
Left: 522 x 450 x 1647 mm, Right: 522 x 450 x 1647 mm | |
Steel | |
Wing | |
1953 mm | |
Q90.195.120.MV | |
Tray shelf | |
1193 mm |
615 mm | |
615 x 1193 x 1953 mm | |
90 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1647 mm | |
2 | |
Powder-coated textured | |
450 mm |