missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet Q-CLASSIC-90 Winged Doors
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3421404133656
3922.77 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 454.25519kg
Alert:
For indoor use only
Description
- Safe and approved storage of flammable hazardous substances in working areas
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Easy handling with comfort: doors stop in any position, self-closing in the event of fire
- No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy transport: integrated transport base with removable, optional base cover
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
- Fire resistance 90 minutes
- Type tested in accordance with EN 14470-1
- EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
1647 mm | |
3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated), 3 x tray shelf, 1 x bottom collecting sump (PP), vertical dividing wall | |
Safe and approved storage of flammable hazardous substances in working areas | |
522 mm | |
Steel | |
615 x 1193 x 1953 mm | |
Left: 522 x 450 x 1647 mm, Right: 522 x 450 x 1647 mm | |
Tray shelf | |
1953 mm | |
Q90.195.120.MV | |
90 mm | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
Light gray doors | |
Safety storage cabinet | |
450 mm | |
Powder-coated textured | |
1193 mm | |
504.50 kg | |
Anthracite Gray, Light Gray | |
615 mm | |
Wing | |
6 | |
2 |