missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet Q-PEGASUS-90 Single Door

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3007404133610

Additional Details : Weight : 309.76199kg

 View more versions of this product

Product Code. 15961916

  • £3404.72 / Each
Estimated Shipment: 26-11-2024
to see stock.

Description

Description

  • Safe and approved storage of flammable hazardous substances in working areas
  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Effortless use: one-hand operation for double-door cabinets (AGT)
  • User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
  • Always closed: serial automatic door closing (TSA) releases arrested doors after approx. 60 seconds; acoustic and visual signal before closing
  • No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
  • Easy transport: integrated transport base with removable, optional base cover
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
Specifications

Specifications

1647 mm
6 x 5 liter drawer (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
522 mm
Steel
615 x 599 x 1953 mm
522 x 450 x 1647 mm
615 mm
Swing, Left-hinged, Auto-close
90 mm
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Light gray doors
Safety storage cabinet
450 mm
Powder-coated textured
599 mm
338.50 kg
Anthracite Gray, Light Gray
1953 mm
Q90.195.060.WDAC
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers