missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet Q-PHOENIX-90
Safe and approved storage of flammable hazardous substances in working areas
£3761.41 - £4846.32
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 615 mm |
Depth (Metric) Interior | 522 mm |
Dimensions (D x W x H) | 615 x 1193 x 1953 mm |
Dimensions (D x W x H) Interior | 522 x 1050 x 1647 mm |
Alert:
For indoor use only
Product Code | Brand | Configuration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Price | Quantity & Availability | |||||
15917446
|
asecos™
3214904133635 |
3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) |
£3761.41
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15907456
|
asecos™
3214904133647 |
4 x drawer (sheet steel powder-coated) |
£4403.24
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927456
|
asecos™
3214904133649 |
6 x drawer (sheet steel powder-coated) |
£4846.32
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Space saving folding doors: less required space in front of the cabinet while doors are open
- Easy handling with comfort: doors stop in any position, self-closing in the event of fire
- No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy transport: integrated transport base with removable, optional base cover
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
522 mm | |
522 x 1050 x 1647 mm | |
Steel | |
Folding | |
1953 mm | |
Q90.195.120.FD | |
Powder-coated textured | |
1050 mm |
615 mm | |
615 x 1193 x 1953 mm | |
90 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1647 mm | |
2 | |
1193 mm |