missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 with Door Open Arrest System - Left-Hinged
Safe and approved storage of flammable hazardous substances in working areas
£2105.69 - £4202.90
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 616 mm |
Depth (Metric) Interior | 524 mm |
Material | Steel |
Product Type | Safety storage cabinet |
Alert:
For indoor use only
Product Code | Brand | Height (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
15903856
|
asecos™
3023900133740 |
1298 mm | 2 x drawer (sheet steel powder-coated) | Light Gray |
£2214.34
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15497524
|
asecos™
3023900130277 |
1298 mm | 2 x shelf, 1 x perforated insert (235 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£2708.30
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15953846
|
asecos™
3023900133735 |
1298 mm | 2 x shelf, 1 x perforated insert (285 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£2105.69
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15913856
|
asecos™
3023900133741 |
1298 mm | 3 x drawer (sheet steel powder-coated) | Light Gray |
£2354.26
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15973876
|
asecos™
3023900233741 |
1298 mm | 3 x drawer (sheet steel powder-coated) | Warning Yellow |
£2354.26
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15963846
|
asecos™
3023900133736 |
1298 mm | 3 x shelf, 1 x perforated insert (185 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£2143.32
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15933876
|
asecos™
3023900233736 |
1298 mm | 3 x shelf, 1 x perforated insert (185 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Yellow |
£2143.32
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13493997
|
asecos™
3022800230249 |
1968 mm | 3 x shelf, 1 x perforated insert (335 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Yellow |
£2824.90
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15943216
|
asecos™
3022800233659 |
1968 mm | 3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow |
£2388.18
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15933186
|
asecos™
3022800133663 |
1968 mm | 3 x tray shelf, 1 x perforated insert (320 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£2589.58
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15983216
|
asecos™
3022800233663 |
1968 mm | 3 x tray shelf, 1 x perforated insert (320 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow |
£2589.58
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15943186
|
asecos™
3022800133681 |
1968 mm | 4 x drawer (sheet steel powder-coated) | Light Gray |
£2739.04
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15963186
|
asecos™
3022800133683 |
1968 mm | 6 x drawer (sheet steel powder-coated) | Light Gray |
£3018.88
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15903226
|
asecos™
3022800233683 |
1968 mm | 6 x drawer (sheet steel powder-coated) | Yellow |
£3018.88
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15296878
|
asecos™
3022800130269 |
1968 mm | 6 x drawer, 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£3593.40
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15135584
|
asecos™
3022800230269 |
1968 mm | 6 x drawer, 1 x bottom collecting sump (sheet steel powder-coated) | Yellow |
£4202.90
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- User-friendly and safe (model .WDAS): door open arrest system, convenient and safe removal or storage of containers
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
524 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1 | |
596 mm |
616 mm | |
Steel | |
Wing, Left-hinged | |
Door open arrest system | |
Powder-coated plain | |
450 mm |