missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 with Door Open Arrest System (Yellow)
Safe and approved storage of flammable hazardous substances in working areas
£2990.79 - £4576.55
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 616 mm |
Depth (Metric) Interior | 524 mm |
Material | Steel |
Product Type | Safety storage cabinet |
Alert:
For indoor use only
Product Code | Brand | Configuration | Height (Metric) Exterior | Width (Metric) Exterior | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Height (Metric) Exterior | Width (Metric) Exterior | Price | Quantity & Availability | |||
15952876
|
asecos™
3012200233753 |
2 x drawer (sheet steel powder-coated) | 1298 mm | 1196 mm |
£3237.24
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15962866
|
asecos™
3012200233744 |
2 x shelf, 1 x perforated insert (285 mm), 1 x bottom collecting sump (sheet steel powder-coated) | 1298 mm | 1196 mm |
£2990.79
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15962876
|
asecos™
3012200233754 |
3 x drawer (sheet steel powder-coated) | 1298 mm | 1196 mm |
£3458.78
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15972866
|
asecos™
3012200233745 |
3 x shelf, 1 x perforated insert (185 mm), 1 x bottom collecting sump (sheet steel powder-coated) | 1298 mm | 1196 mm |
£3038.49
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15923036
|
asecos™
3020500233705 |
3 x shelf, 1 x perforated insert (370 mm), 1 x bottom collecting sump (sheet steel powder-coated) | 1968 mm | 896 mm |
£2990.79
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13420323
|
asecos™
3011600230127 |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | 1968 mm | 1196 mm |
£3763.00
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15902576
|
asecos™
3011600233715 |
3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) | 1968 mm | 1196 mm |
£3093.61
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15974346
|
asecos™
3034800233848 |
3 x shelf, 1 x perforated insert (370 mm), 1 x bottom collecting sump (sheet steel powder-coated), 3 x tray shelf, 1 x bottom collecting sump (PP), vertical dividing wall | 1968 mm | 1196 mm |
£3945.85
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15963036
|
asecos™
3020500233709 |
4 x drawer (sheet steel powder-coated) | 1968 mm | 896 mm |
£3632.62
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15982576
|
asecos™
3011600233726 |
6 x drawer (sheet steel powder-coated) | 1968 mm | 1196 mm |
£4178.52
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15984346
|
asecos™
3034800233849 |
6 x drawer (sheet steel powder-coated), 3 x tray shelf, 1 x bottom collecting sump (PP), vertical dividing wall | 1968 mm | 1196 mm |
£4576.55
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- User-friendly and safe (model .WDAS): door open arrest system, convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
524 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
2 |
616 mm | |
Steel | |
Wing | |
Door open arrest system | |
Powder-coated plain |