missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 Left-Hinged

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3022700133663

Additional Details : Weight : 281.66849kg

 View more versions of this product

Product Code. 15973116

  • £2501.60 / Each
Estimated Shipment: 26-11-2024
to see stock.

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications

Specifications

1742 mm
3 x tray shelf, 1 x perforated insert (320 mm), 1 x bottom collecting sump (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
524 mm
Steel
616 x 596 x 1968 mm
524 x 450 x 1742 mm
Tray shelf
1968 mm
S90.196.060
120 mm
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Tall
Safety storage cabinet
450 mm
Powder-coated plain
596 mm
284 kg
Light Gray
616 mm
Swing, Left-hinged
3
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers