missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 Right-Hinged
Safe and approved storage of flammable hazardous substances in working areas
£2017.71 - £2501.60
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 616 mm |
Depth (Metric) Interior | 524 mm |
Material | Steel |
Product Type | Safety storage cabinet |
Alert:
For indoor use only
Product Code | Brand | Height (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
15973906
|
asecos™
3024000134413 |
1298 mm | 2 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£2017.71
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15933946
|
asecos™
3024000234413 |
1298 mm | 2 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow |
£2017.71
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15983906
|
asecos™
3024000134414 |
1298 mm | 3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£2055.34
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15943946
|
asecos™
3024000234414 |
1298 mm | 3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow |
£2055.34
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15973256
|
asecos™
3022900134387 |
1968 mm | 3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£2300.20
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15913306
|
asecos™
3022900234387 |
1968 mm | 3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Yellow |
£2300.20
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15913266
|
asecos™
3022900134391 |
1968 mm | 3 x tray shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£2501.60
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15953306
|
asecos™
3022900234391 |
1968 mm | 3 x tray shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow |
£2501.60
Each |
Estimated Shipment: 08-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
524 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
Powder-coated plain | |
450 mm |
616 mm | |
Steel | |
Swing, Right-hinged | |
1 | |
596 mm |