missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90
Safe and approved storage of flammable hazardous substances in working areas
£2819.07 - £3768.30
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 616 mm |
Depth (Metric) Interior | 524 mm |
Material | Steel |
Product Type | Safety storage cabinet |
Product Code | Brand | Configuration | Color | Height (Metric) Exterior | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Color | Height (Metric) Exterior | Price | Quantity & Availability | |||
15155155
|
asecos™
3012100230155 |
2 x shelf, 1 x perforated insert (275 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Yellow | 1298 mm |
£2835.50
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15922786
|
asecos™
3012100133744 |
2 x shelf, 1 x perforated insert (285 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray | 1298 mm |
£2819.07
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15566855
|
asecos™
3012100233744 |
2 x shelf, 1 x perforated insert (285 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Yellow | 1298 mm |
£2819.07
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15932956
|
asecos™
3020400133705 |
3 x shelf, 1 x perforated insert (370 mm), 1 x 22 liter bottom collecting sump (sheet steel powder-coated) | Light Gray | 1968 mm |
£2819.07
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15902986
|
asecos™
3020400233705 |
3 x shelf, 1 x perforated insert (370 mm), 1 x 22 liter bottom collecting sump (sheet steel powder-coated) | Yellow | 1968 mm |
£2819.07
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15932786
|
asecos™
3012100133745 |
3 x shelf, 1 x perforated insert (185 mm), 1 x 33 liter bottom collecting sump (sheet steel powder-coated) | Light Gray | 1298 mm |
£2866.77
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15902816
|
asecos™
3012100233745 |
3 x shelf, 1 x perforated insert (185 mm), 1 x 33 liter bottom collecting sump (sheet steel powder-coated) | Yellow | 1298 mm |
£2866.77
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15536855
|
asecos™
3011500233715 |
3 x shelf, 1 x perforated insert (380 mm), 1 x 33 litre bottom collecting sump (sheet steel powder-coated) | Warning Yellow | 1968 mm |
£2921.89
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15317438
|
asecos™
3011500130133 |
3 x tray shelf, 1 x perforated insert (111 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray | 1968 mm |
£3768.30
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15992476
|
asecos™
3011500133719 |
3 x tray shelf, 1 x perforated insert (135 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray | 1968 mm |
£3124.35
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15992506
|
asecos™
3011500233719 |
3 x tray shelf, 1 x perforated insert (135 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow | 1968 mm |
£3124.35
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
524 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
3 |
616 mm | |
Steel | |
Wing | |
2 | |
Powder-coated plain |