missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PHOENIX-90 Left-Hinged
Safe and approved storage of flammable hazardous substances in working areas
£2486.76 - £3287.06
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Material | Steel |
Product Type | Safety storage cabinet |
Door Style | Folding, Left-hinged |
For Use With (Application) | Safe and approved storage of flammable hazardous substances in working areas |
Alert:
For indoor use only
Product Code | Brand | Depth (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Depth (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
15943426
|
asecos™
3023300133659 |
616 mm | 3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump | Light Gray |
£2486.76
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15903466
|
asecos™
3023300233659 |
616 mm | 3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump | Yellow |
£2486.76
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15983426
|
asecos™
3023300133663 |
616 mm | 3 x tray shelf, 1 x perforated insert (320 mm), 1 x bottom collecting sump | Light Gray |
£2688.16
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15943466
|
asecos™
3023300233663 |
616 mm | 3 x tray shelf, 1 x perforated insert (320 mm), 1 x bottom collecting sump | Warning Yellow |
£2688.16
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15993426
|
asecos™
3023300133681 |
616 mm | 4 x drawer | Light Gray |
£2837.62
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15953466
|
asecos™
3023300233681 |
616 mm | 4 x drawer | Warning Yellow |
£2837.62
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15913436
|
asecos™
3023300133683 |
616 mm | 6 x drawer | Light Gray |
£3117.46
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15973466
|
asecos™
3023300233683 |
616 mm | 6 x drawer | Yellow |
£3117.46
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15914166
|
asecos™
3031300133695 |
749 mm | 4 x drawer | Light Gray |
£2966.94
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15974176
|
asecos™
3031300233695 |
749 mm | 4 x drawer | Yellow |
£2966.94
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15934166
|
asecos™
3031300133697 |
749 mm | 6 x drawer | Light Gray |
£3287.06
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15994176
|
asecos™
3031300233697 |
749 mm | 6 x drawer | Yellow |
£3287.06
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Space saving folding door: less required space in front of the cabinet while door is open
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1742 mm | |
1 | |
596 mm |
Steel | |
Folding, Left-hinged | |
1968 mm | |
Door open arrest system | |
Powder-coated plain | |
450 mm |