missing translation for 'onlineSavingsMsg'
Learn More

asecos Type 90 Safety Storage Cabinet V-CLASSIC-90

The MULTIRISK cabinet from asecos for safe, flexible, and energetically sustainable storage of various hazardous materials in just one cabinet.

Brand:  asecos 4028500540498

Additional Details : Weight : 463.00000kg

 View more versions of this product

Product Code. 17815814

  • £7250.40 / Each
Estimated Shipment: 22-11-2024
to see stock.

Alert:

For indoor use only


Description

Description

Function / construction

  • Special fire resistant construction: The special, completely fire-resistant construction of the Type 90 MULTIRISK cabinet offers, for the first time, the possibility of fire-protected storage of corrosive and toxic hazardous substances
  • The cabinet thus not only ensures the safe storage of different hazardous materials in daily operations, but also provides maximum protection for users and rescue forces in case of a fire
  • Locking: To protect against unauthorised use, the cabinet can be securely locked
  • The integrated red/green indicator quickly shows the locking status
  • The cabinet can be integrated into existing locking systems by replacing the profile cylinder on site
  • Comfortable overview and access: The cabinet user is entirely unrestricted in deciding which types of hazardous materials are to be stored in the two storage compartments
  • Both vertical drawers can be used equally thanks to the ventilation concept and uniform interior equipment
  • Metal-free storage levels: Each storage level is equipped with an tray shelf made of chemical-resistant PE and can be adjusted tool-free in height at any time by the customer
  • The polyethylene trays are easy to remove, e.g. for cleaning or in case of leakage
  • Optimal integration and use of space: The space-saving storage solution with high storage volume is optimally suited for installation next to fume cupboards with a width of only 60 cm and a depth of 86 cm deep
  • Technical ventilation: Hazardous material vapours are captured at each storage level within the cabinet and can be extracted directly via the integrated air ducts ready for conneciton (NW 75) to a technical exhaust system
  • The possible formation of explosive atmospheres or harmful vapours is safely prevented

Certificates

  • Fire resistance 90 minutes
  • Type tested in accordance with EN 14470-1
  • DIN EN 16121/16122 conformity
  • GS certified
  • CE compliant
Specifications

Specifications

688.98 in.
For the unrestricted storage of a variety of hazardous materials in work areas
236.22 in.
Per Door: 1 x tray shelf with storage box (powder-coated sheet steel), 3 x tray shelf metal free, 4 x perforated insert (PP), 1 x bottom collecting sump (PE)
463 kg
306.69 in.
White
1020.74 lb.
Vertical Drawer
8
2
199.21 in.
Safety Storage Cabinet
774.02 in.
Gypsum
860 x 600 x 1966 mm
338,583 in.
779 x 506 x 1750 mm
Tray Shelf, Tray Shelf with Storage Box
30 kg
V90.196.060.MV.2VD:0012
1750 mm
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers