missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet XL-CLASSIC-90
Safe and approved storage of flammable hazardous substances in working areas
£5170.68 - £6340.92
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 1018 mm |
Depth (Metric) Interior | 864 mm |
Color | Light Gray |
Material | Steel |
Alert:
For indoor use only
Product Code | Brand | Width (Metric) Interior | Configuration | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) Interior | Configuration | Price | Quantity & Availability | ||||
15974386
|
asecos™
3039300130397 |
936 mm | 1 x shelf (sheet steel powder-coated), 1 x transport base (steel powder-coated), 1 x bottom collecting sump with grid (sheet steel galvanized) |
£5300.00
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15954386
|
asecos™
3039300130395 |
936 mm | 1 x transport base (steel powder-coated), 1 x bottom collecting sump with grid (sheet steel galvanized) |
£5170.68
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15944376
|
asecos™
3038200130384 |
1386 mm | 1 x transport base (steel powder-coated), 1 x bottom collecting sump with grid (sheet steel galvanized) |
£6170.26
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15964376
|
asecos™
3038200130388 |
1386 mm | 1 x bottom collecting sump with shelves, 1 x transport base (steel powder-coated) |
£6340.92
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with cylinder lock
- Internal flexibility: transport base (optional) for safe and easy in-house transport
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Safety while decanting: end-to-end earthing link at the rear panel, inside with earthing wires incl. clamp
- Certificates
- Fire resistance 90 minutes
- Type tested in accordance with EN 14470-1
- GS certified
- CE compliant
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, GS certified, CE compliant | |
864 mm | |
Steel | |
Wing | |
2224 mm | |
Door open arrest system | |
Powder-coated plain |
1018 mm | |
Light Gray | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1890 mm | |
2 |