missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Under Bench Cabinet UB-T-90
Safe and approved storage of flammable hazardous substances in working areas
£1957.82 - £3080.36
Specifications
Certifications/Compliance | EN 14470-1 with a fire resistance of 90 minutes |
---|---|
Depth (Metric) Interior | 482 mm |
Color | Light Gray |
Material | Steel |
For Use With (Application) | Storage of flammable liquids |
Alert:
For indoor use only
Product Code | Brand | Dimensions (D x W x H) Interior | Configuration | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (D x W x H) Interior | Configuration | Price | Quantity & Availability | ||||
15365569
|
asecos™
3265700135373 |
482 x 979 x 702 mm | 2 drawers, 2 perforated inserts |
£3080.36
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15365579
|
asecos™
3265800135378 |
482 x 470 x 702 mm | One pullout drawer, perforated metal plate insert and one second level drawer |
£1957.82
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Safe and approved storage of flammable hazardous substances in working areas
Function / construction
- Robust construction and longevity: body made of powder-coated sheet steel
- Easy handling with comfort: doors stop in any position, self-closing in the event of fire
- User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
- No unauthorised use: doors lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
Certificates
- Fire resistance 90 minutes
- type tested in accordance with EN 14470-1
- EN 16121/16122 conformity
- GS certified
- CE compliant
Compliance
DIN EN 14470-1 - Fire resistance of 90 minutes
Specifications
EN 14470-1 with a fire resistance of 90 minutes | |
Light Gray | |
Storage of flammable liquids | |
702 mm |
482 mm | |
Steel | |
800 mm | |
Powder-coated |