missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Type B Safety Bumper

Protect corners and edges with this Type B Safety Bumper. Dual function: indicates hazardous situations and softens any impacts.

Brand:  Brady™ 225023

Code : NEW

Additional Details : Weight : 2.00000kg

Product Code. 11716815

  • £141.00 / 1 pack
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant against hydrocarbons.
Specifications

Specifications

10 Pk.
Self-adhesive Safety Bumpers - Sleeve (b)
Safety Identification
Safety Bumpers
Cellular Thermoplastic
25.98 in.
Black and Yellow Striped
28 mm
1.1 in.
0.66 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers