missing translation for 'onlineSavingsMsg'
Learn More

Testo™ 925 Single Channel Thermometer

Use this thermometer for heating engineering, the sanitation sector and refrigeration and air conditioning technology applications. Testo™ 925 Single Channel Thermometers enable virtually any kind of temperature measurement.

Brand:  Testo™ 0560 9250

Code : A0

Additional Details : Weight : 0.33300kg

Product Code. 10185252

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • An attachable temperature probe and a radio temperature probe can be connected
  • Large range of fast temperature probes on offer for surface, air or immersion/penetration measurement (optional)
  • Continuous display of min./max. values
  • Acoustic alarm sounds when limit values are exceeded
Specifications

Specifications

Testo 925, 1 channel temperature measuring instrument T/C Type K, audible alarm, connection of an optional radio probe, Battery, calibration protocol
Type K (NiCr-Ni)
Thermocouple Thermometer
-20°C
±(0.5°C 0.3% of mv), (−40 to 900°C), ±(0.7°C 0.5% of mv)
ABS Plastic
0.1°C (−50 to 199.9°C), 1°C
50°C
-20°C to +50°C
Large
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers