Learn More
TYRO3 Monoclonal Antibody (10E11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515505
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15505 targets TYRO3 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15505 immunogen is purified recombinant extracellular fragment of human TYRO3 fused with hIgGFc tag expressed in HEK293 cell line. MA5-15505 detects TYRO3 which has a predicted molecular weight of approximately 96.9kDa.
TYRO3 (tyrosine-protein kinase receptor TYRO3) is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding to several ligands including TULP1 or GAS6. It regulates many physiological processes including cell survival, migration, and differentiation. Ligand binding at the cell surface induces dimerization and autophosphorylation of TYRO3 on its intracellular domain that provides docking sites for downstream signaling molecules. Following activation by a ligand, TYRO3 plays a role in various processes such as neuron protection from excitotoxic injury, platelet aggregation, signal transduction, cell adhesion. and cytoskeleton reorganization.Specifications
TYRO3 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q06418 | |
TYRO3 | |
Purified recombinant extracellular fragment of human TYRO3 fused with hIgGFc tag expressed in HEK293 cell line. | |
RUO | |
7301 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
10E11 | |
Unconjugated | |
TYRO3 | |
AI323366; Brt; Bruton agammaglobulinemia tyrosine kinase; BYK; Dtk; Etk-2; etk2/tyro3; FLJ16467; Rek; Rse; Sky; soluble tyro 3; soluble tyro3; styro 3; styro3; TIF; TK19-2; tyro 3; TYRO3; TYRO3 protein tyrosine kinase; TYRO3 protein tyrosine kinase 3; tyrosine-protein kinase byk; Tyrosine-protein kinase DTK; tyrosine-protein kinase receptor TYRO3; Tyrosine-protein kinase RSE; tyrosine-protein kinase SKY; Tyrosine-protein kinase TIF | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |