missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Tyrosine Hydroxylase Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Brand: Novus Biologicals™ NBP2-46647PEP
Additional Details : Weight : 0.00970kg
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human TH. The Tyrosine Hydroxylase Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.Specifications
7054 | |
DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase | |
PBS (pH 7.4), 1 M Urea | |
25 kDa | |
Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Prostate Cancer | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-53937. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
>80% by SDS-PAGE and Coomassie blue staining | |
Store at -20°C. Avoid freeze-thaw cycles. | |
TH | |
100 μL | |
E. coli |