missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tyrphostin AG-213, 97%

£72.33 - £311.00

Specifications

Description Orange
Content And Storage Ambient temperatures
Assay 97%
CAS 122520-86-9
Molecular Formula C10H8N2O2S
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15458709
View Documents
Thermo Scientific™
J67345.#0
5 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15468709
View Documents
Thermo Scientific™
J67345.MA
10 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15478709
View Documents
Thermo Scientific™
J67345.MB
25 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Orange
97%
C10H8N2O2S
RG-50864; Tyrphostin 47
Ambient temperatures
122520-86-9
220.25g/mol
Solid
Videos
SDS
Documents

Documents

Product Certifications