missing translation for 'onlineSavingsMsg'
Learn More
DuPont™ Tyvek™ 500 Lab Coats Product Code.: 16387381
Add to basket

DuPont™ Tyvek™ 500 Lab Coats

Product Code. 16387381 Shop All DuPont Products
Large
Click to view available options
Size:
Small
Medium
Large
2X-Large
This item is not returnable. View return policy

Product Code. 16387381

Brand: DuPont™ D15534012

Add to basket
Add to basket
This item is not returnable. View return policy

Tyvek™ Lab coats offer enhanced protection for body parts that are more exposed to hazardous substances.

  • Composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort
  • Permeable to both air and water vapour
  • Offers an excellent barrier against fine particles and fibres (down to 1 micron in size)
  • Certified according to Regulation (EU) 2016/425
  • Partial body chemical protective clothing, Category III, Type PB (6-B)
  • EN 14126 (barrier to infective agents)
  • Antistatic treatment (EN 1149-5) on both sides
  • Applications include: pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting, and general clean-up
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Resistant Lab Coat
Size Large
Gender Unisex
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Limited-use
Sterility Non-sterile
Resistance Chemicals
Seam Type Stitched
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tyvek™ 500 Lab Coats >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.