missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 500 Lab Coats

Product Code. 16397381
This item is not returnable. View return policy
This item is not returnable. View return policy

Lab coats offer enhanced protection for body parts that are more exposed to hazardous substances.

  • Composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort
  • Permeable to both air and water vapour
  • Offers an excellent barrier against fine particles and fibres (down to 1 micron in size)
  • Certified according to Regulation (EU) 2016/425
  • Partial body chemical protective clothing, Category III, Type PB (6-B)
  • EN 14126 (barrier to infective agents)
  • Antistatic treatment (EN 1149-5) on both sides
  • Applications include: pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting, and general clean-up
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Resistant Lab Coat
Gender Unisex
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Limited-use
Sterility Non-sterile
Resistance Chemicals
Seam Type Stitched
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'