missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek 600 Plus White Hooded Coverall
Providing an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ Taped Seam Hooded Coveralls are composed of flash spun high density polyethylene.
£225.00 - £860.00
Specifications
Product Type | Hooded Coveralls |
---|---|
Material | HDPE |
Color | White |
Ankle Style | Elastic |
Cuff Style | Thumb Loop |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11703083
|
DuPont™
D13395307 |
Medium |
£834.00
Pack of 100 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
12372239
|
DuPont™
D13395307 |
Medium |
£225.00
Pack of 25 |
In Stock
Log in to see stock. |
|||||
11723083
|
DuPont™
D13395284 |
X-Large |
£834.00
Pack of 100 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
12392239
|
DuPont™
D13395284 |
X-Large |
£225.00
Pack of 25 |
In Stock
Log in to see stock. |
|||||
11743083
|
DuPont™
D13495752 |
3X-Large |
£860.00
Pack of 100 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
12312249
|
DuPont™
D13495752 |
3X-Large |
£229.00
Pack of 25 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
12362239
|
DuPont™
D13495782 |
Small |
£225.00
Pack of 25 |
In Stock
Log in to see stock. |
|||||
11793073
|
DuPont™
D13495782 |
Small |
£834.00
Pack of 100 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
11713083
|
DuPont™
D13395299 |
Large |
£834.00
Pack of 100 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
12382239
|
DuPont™
D13395299 |
Large |
£225.00
Pack of 25 |
In Stock
Log in to see stock. |
|||||
11733083
|
DuPont™
D13395272 |
2X-Large |
£819.00
Pack of 100 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
12302249
|
DuPont™
D13395272 |
2X-Large |
£225.00
Pack of 25 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
Description
- Self-adhesive chin strap for tight seal of suit to face mask
- Stitched and over-taped seams for protection and strength
- Tyvek zipper and zipper flap for enhanced protection
- Elasticated face, wrists and ankles as well as glued-in elastic waist
- Elasticated thumb loops prevent sleeves from riding up
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Applications include maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or research, biosecurity laboratories as well as medical applications and when exposed to biological hazards
Specifications
Hooded Coveralls | |
White | |
Thumb Loop | |
Elastic | |
Stitched, Taped | |
Chemicals, Liquid | |
Maintenance and Dismantling Jobs in the Nuclear Industry, Pharmaceutical Manufacturing or in Research and Biosecurity Laboratories |
HDPE | |
Elastic | |
Hooded | |
Zipper with Self-adhesive Flap and Chin Flap | |
Category III, Type 4-B, 5-B and 6-B | |
Category III, Type 4-B, 5-B and 6-B, EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), (EN 1149-5) |