missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem 6000 F Sleeves
135.59 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Length: 500mm
Description
- Antistatic treatment
- Wide elastics at cuff and upper arm
- Excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents
- Used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
Specifications
Size | Universal |
Construction | Heavy-duty Tyvek substrate |
Product Type | Chemical-resistant Sleeve |
Material | Tyvek Substrate |
Color | Gray |
Seam Type | Stitched, Over-taped |
Cuff Style | Elastic |
Finish | Polymeric barrier film |
Certifications/Compliance | (EU) 2016/425, Category III, Type PB [3], EN 14126, EN 1149-5 |
Length (Metric) | 500 mm |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'