missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 600 Plus with Socks

Product Code. 11833510 Shop All DuPont Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
Pack of 20
Pack of 80

Product Code. 11833510

Brand: DuPont™ D13398948

Only null left
Add to basket
Add to basket

Composed of flash spun high density polyethylene. Dupont™ Tyvek™ 600 Plus with Socks is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.

  • Hooded coverall with integrated socks and additional knee-length boot flap.
  • Available in white in sizes S to XXXL.
  • Robust yet lightweight (<250g per garment).
  • Self-adhesive chin flap for tight seal of suit to the mask.
  • Elasticated face, wrists and ankles as well as glued-in waist elastic.
  • Elasticated thumb loops prevent sleeves from riding up.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hooded Coveralls
Size Large
Material HDPE
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic, Attached Socks, Boot Overflaps
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper with Self-adhesive Flap and Chin Flap
Seam Type Stitched, Taped
Protection Class Category III, Type 4-B, 5-B and 6-B
Collar Type Self-adhesive chin flap
Certifications/Compliance (EU) 2016/425, EN 14126, EN 1149-5
Size (Metric) Chest 100 to 108 cm
For Use With (Application) For maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or in research and biosecurity laboratories, medical applications and when exposed to biological hazards
Height (Metric) 174 to 182 cm
Show More Show Less

The user should make the risk assessment to determine the protection required.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.