Learn More
DuPont™ Tyvek 500 Jacket Product Code.: 12633690
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
DuPont™ Tyvek 500 Jacket
Product Code. 12633690 Shop All DuPont Products
X-Large
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Product Code. 12633690
Brand: DuPont™ D13396450
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
Add to basket
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ 500 Jacket with Hood is composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
  • Specially designed for use with Tyvek™ apparel
  • Flash-spun HDPE construction
  • Stitched internal seams
  • Front zip closure
  • Elasticated face and wrists
  • White
  • PPE Category I
  • Antistatic treatment (EN 1149-5) - on both sides
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Material HDPE
Color White
Closure Type Zipper
Certifications/Compliance EN 1149-5
Seam Type Stitched (Internal)
Product Type Hooded Chemical-resistant Jacket
For Use With (Application) Pharmaceutical Handling, Chemical Protection, Lead and Asbestos Abatement/remediation, General Maintenance/operations, Spray Painting and General Clean-up, Amongst Many Others
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
  • Multiple Manufacturers
    Discover exclusive monthly deals on essential safety products to enhance your lab's protection and compliance.
    Promo Code: 24259
    Expires2025-04-30
Product Content Correction
Product Title
DuPont™ Tyvek 500 Jacket >