Learn More
DuPont™ Tyvek 500 Industry Coveralls
GREENER_CHOICE
Product Code. 12932219
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
Pack of 100
Pack of 25
Product Code. 12932219
Brand: DuPont™ D13984290
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ 500 Industry Coveralls with Collar are composed of flash spun high density polyethylene.
  • With collar
  • Internal stitched seams
  • Zipper made of TYVEK™ with flap
  • Elastic cuffs, ankles and waist
  • Antistatic treatment
  • Category III, Type 5/6
Order Info
dimSize
TRUSTED_SUSTAINABILITY
Specifications
Product Type Collared Coveralls
Size 3X-Large
Material HDPE
Color White
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Zipper with Storm Flap
Seam Type Stitched, Taped
Protection Class Category III, Type 5 and 6
Collar Type Standard fold
Certifications/Compliance EN 1073-2, EN 1149-5
Description Zipper flap, tunneled cuff and facial elastics
Size (Metric) Chest 124 to 132 cm
Quantity 100/Pk.