missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek 500 Industry Coveralls
GREENER_CHOICE

Product Code. 15381521
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
Pack of 100
Pack of 25

Product Code. 15381521

Brand: DuPont™ D13395359

Only null left
Add to basket
Edge
Add to basket

Provides an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ 500 Industry Coveralls with Collar are composed of flash spun high density polyethylene.

  • With collar
  • Internal stitched seams
  • Zipper made of TYVEK™ with flap
  • Elastic cuffs, ankles and waist
  • Antistatic treatment
  • Category III, Type 5/6
TRUSTED_SUSTAINABILITY

Specifications

Product Type Collared Coveralls
Size Medium
Material HDPE
Color White
Waist Style Elastic (Stitched-in)
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Zipper with Storm Flap
Seam Type Stitched
Protection Class Category III, Type 5 and 6
Collar Type Standard fold
Certifications/Compliance EN 1073-2, EN 1149-5
Size (Metric) Chest 92 to 100 cm
For Use With (Application) For industrial applications
Height (Metric) 168 to 176 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.