Learn More
DuPont™ Tyvek™ IsoClean™ Boot Covers, Model IC 458
Shop All DuPont Products
Click to view available options
Size:
Medium
Large
Description
- Gripper™ soles provide enhanced slip-retardance and robustness.
- Delivers an ideal balance of protection, durability and comfort.
- Suitable for use in GMP class C/D (ISO Class 6-9) clean rooms.
Specifications
Specifications
Size | Medium |
Material | HDPE |
Color | White |
Sterility | Non-sterile |
Closure Type | Ties |
Sole Type | Slip-resistant |
Seam Type | Serged |
Certifications/Compliance (Europe) | (EU) 2016/425, EN 14126 |
Product Type | Boot Covers |