missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Bouffant Cap
220.34 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Product Type: Bouffant Cap
Description
- Elastic headband
- One size fits most
- Tyvek™ IsoClean™ (option codes 0B, 00 and BH) garments and accessories are neither clean-processed nor gamma-irradiated but manufactured in a controlled environment.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Product Type | Cleanroom Bouffant |
Size | One Size |
Color | White |
Model | D15531628 |
Seam Type | Serged |
Sterility | Non-sterile |
For Use With (Application) | Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Cosmetics Industry |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'