Learn More
DuPont™ Tyvek™ IsoClean™ Chemo Gown Product Code.: 17876865
Estimated Shipment: 28-07-2025
to see stock.
Add to basket
DuPont™ Tyvek™ IsoClean™ Chemo Gown
Product Code. 17876865
X-Small
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Product Code. 17876865
Brand: DuPont™ D15542351
Estimated Shipment: 28-07-2025
to see stock.
Add to basket
Add to basket
Collared gown with hook and loop closure in the neck, serged seams, knit cuffs and bound ties at waist originating from elasticated sides. Not clean-processed and not sterilized.
  • Available in sizes XS to 3X
  • Increased skirt length ensures protection against frontal exposure Openness in the back closure increases wearer comfort
  • Delivers an ideal balance of protection, durability and comfort
  • Made of high density polyethylene using a patented flash spinning process
  • Provides an inherent barrier to particles, microorganisms and non-hazardous water-based liquid splash
  • Manufactured in a controlled environment
TRUSTED_SUSTAINABILITY
Specifications
Size X-Small
Product Type Controlled Environments Gown
Material Polyethylene
Color White
Closure Type Hook and Loop
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Latex Free Yes
Sleeve Style Long Sleeve
Autoclavable Autoclavable
Product Title
DuPont™ Tyvek™ IsoClean™ Chemo Gown >