missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Cleanroom Sleeve
Shop All DuPont Products
Click to view available options
Type: Sleeve
Description
- Tunnelled elastication at wrist and bicep
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash
- Breathable and exceptionally low linting
- Accessory

Specifications
Specifications
Product Type | Controlled Environment Sleeve |
Material | Polyethylene |
Sterility | Non-sterile |
Seam Type | Bound |
Model | IC501B00 |
For Use With (Application) | Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Other Controlled Environments |
Certifications/Compliance (Europe) | (EU) 2016/425, EN 14126 |
Length (Metric) | 45 cm |
Cleanroom Class | Class 1000-100,000 (ISO 6-8) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DuPont™ Tyvek™ IsoClean™ Cleanroom Sleeve >
Spot an opportunity for improvement?Share a Content Correction