missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck
142.82 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Size:
Small
Medium
X-Large
2X-Large
3X-Large
Description
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash
- Breathable and exceptionally low linting
Specifications
Product Type | Controlled Environments Lab Coat |
Size | 2X-Large |
Material | Polyethylene |
Color | White |
Closure Type | Snaps |
Sterility | Non-sterile |
Model | IC270B0 |
Seam Type | Bound |
Certifications/Compliance (Europe) | (EU) 2016/425, EN 14126 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'