Learn More
DuPont™ Tyvek™ IsoClean™ IC 458B WS Boot Cover, Sterile, Double Bagged
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Description
- Gamma-sterilized
- Bound internal seams
- Tunnelled elastication at shin and ankles
- Ankle ties
- Gripper™ soles provide enhanced slip-retardance and robustness
Tyvek™ IsoClean™ delivers an ideal balance of protection, durability and comfort. Made of high density polyethylene using a patented flash spinning process Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Tyvek™ IsoClean™ is also breathable and low linting.
Tyvek™ IsoClean™ accessories (option code WS) have been sterilized by gamma- irradiation. They are folded to aid aseptic donning and packaged in an ISO class 5 cleanroom. All DuPont™ Tyvek™ IsoClean™ sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag. The packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas. The accessories are individually packed and grouped together in an outer bag.
Garments and accessories made of sterile Tyvek™ IsoClean™ are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Size | X-Large |
Material | Tyvek |
Latex Free | Yes |
Color | White |
Disposable | Single-use |
Sterility | Sterile |
Closure Type | Elastic Top with Ankle Ties |
Sole Type | Anti-skid |
Resistance | Dry Particulates, Splash |
Product Type | Controlled Environments Boot Covers |