Learn More
DuPont™ Tyvek™ IsoClean™ IC 689B WS Hood with Integrated Mask, Sterile, Double Bagged
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Use promo code "22918" to get your promotional price.
Description
- With attached mask and ties for adjustable fit in the chest area
- Available in white and in one size
- Gamma-sterilized and double bagged
- Bound internal seams
- Bound face opening for low particle shedding
Tyvek™ IsoClean™ delivers an ideal balance of protection, durability and comfort. Made of high density polyethylene using a patented flash spinning process. Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash. Tyvek™ IsoClean™ is also breathable and low linting.
Tyvek™ IsoClean™ WS accessories have been sterilized by gamma-irradiation. They are folded to aid aseptic donning and packaged in an ISO class 5 cleanroom. All DuPont™ Tyvek™ IsoClean™ sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag. The packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas. The accessories are individually double packed.
Garments and accessories made of sterile Tyvek™ IsoClean™ are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Product Type | Cleanroom Face Mask |
Color | White |
Fastening Type | Ties |
Material | Tyvek |
Size | One Size |
Sterility | Sterile |
Style | Pleated |
Cleanroom Class | Class 100 (ISO 5) |
Disposable | Single-use |
Latex Free | Yes |