missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ IC193B TS Coverall, Sterile, Double Bagged
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
268.81 GBP valid until 2024-12-31
Use promo code "22918" to get your promotional price.
Use promo code "22918" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Gamma-sterilized
- Bound internal seams
- Elasticated waist at back for good fit
- Tyvek™ covered elasticated thumb loops
- Tunneled elastication at wrists and ankles
- Front zipper closure
- Storm flap
- Delivers an ideal balance of protection, durability and comfort
- Made of high density polyethylene using a patented flash spinning process
- Provides an inherent barrier to particles, microorganisms, and non-hazardous water-based light liquid splash
- Breathable and low linting
- Sterilized by gamma-irradiation
- Folded to aid aseptic donning and packaged in an ISO class 5 cleanroom
- Packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag
- Packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas
- Typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Product Type | Cleanroom Coveralls |
Size | Small |
Gender | Unisex |
Material | Tyvek™ |
Color | White |
Disposable | Single-use |
Waist Style | Elastic |
Ankle Style | Attached Boots |
Cuff Style | Finger Loops |
Neck Style | Hooded |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'